Cesium chloride 99.9%, powder, Optical Grade

Supplier: 0000012207

04813700 04813711 04813729
IC813700EA 2878.47 USD
IC813700 IC813711 IC813729
Cesium chloride 99.9%, powder, Optical Grade
Cesium chloride

Cesium chloride is used for the preparation of electrically conducting glasses.


Used for density gradient work and for the purification of virus/phage, nucleic acids and nucleoproteins


Cesium chloride is used for the preparation of electrically conducting glasses. Used to make solutions for the separation of RNA and DNA, virus/phage by density gradient centrifugation.


Freely soluble in water (50% aqueous - clear, colorless solution); soluble in ethanol, alcohol.

Formula: CsCl
Boiling Pt: 1303 °C (1013 hPa)
Melting Pt: 645 °C (lit.)
Density: 3.7808 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010955
CAS Number: 7647-17-8
EINECS: 231-600-2
Merck Index: 13,02021

Order Now

Specification Test Results

Purity 99.9%
pH (1% aq soln) 5.5 - 6.5
Barium ≤10 ppm
Magnesium ≤10 ppm
Potassium ≤30 ppm
Iron ≤10 ppm
Calcium ≤10 ppm
Solubility (50% aq soln) Clear and Colorless


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR