Glycine ≥99.5%, white crystalline powder for electrophoresis

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: Aminoacetic acid, Glycocoll, H-Gly-OH

04808822 04808831
IC808822EA 144.42 USD
IC808822 IC808831
Glycine ≥99.5%, white crystalline powder for electrophoresis
Glycine

Glycine is an non-essential amino acid with no asymmetric carbon and major inhibitory neurotransmitter. It can fit into hydrophilic or hydrophobic environments, due to its minimal side chain of only one hydrogen atom. It contains heavy metals ≤ 20 ppm. It is commonly used as a component in Tris-glycine and Tris-glycine-SDS running buffers for polyacrylamide gel electrophoresis, a component of Towbin's transfer buffer for Western blots, a buffer substance in cryoenzymology, in osmotic pressure maintenance in isoelectric focusing of erythrocytes, salting-in effect in protein chemistry, and as a buffer component in the coupled phosphatase-kinase reaction for end labelling of restriction fragments.
Glycine is a component of Tris-glycine and Tris-glycine-SDS running buffers for polyacrylamide gel electrophoresis. Glycine is also a component of Towbin's transfer buffer for Western blots.
Inhibitory neurotransmitter in spinal cord, allosteric regulator of NMDA receptors.


  • Electrophoresis Grade
  • Presentation: White Crystalline Powder
  • pKa at 25°C=2.35
  • Soluble in water, slightly soluble in alcohol and ethanol(95%); almost insoluble in ether.
  • Room Temperature


Glycine is an non-essential amino acid with no asymmetric carbon and major inhibitory neurotransmitter. It can fit into hydrophilic or hydrophobic environments, due to its minimal side chain of only one hydrogen atom. It contains heavy metals ≤ 20 ppm. It is commonly used as a component in Tris-glycine and Tris-glycine-SDS running buffers for polyacrylamide gel electrophoresis, a component of Towbin's transfer buffer for Western blots, a buffer substance in cryoenzymology, in osmotic pressure maintenance in isoelectric focusing of erythrocytes, salting-in effect in protein chemistry, and as a buffer component in the coupled phosphatase-kinase reaction for end labelling of restriction fragments.


Glycine is a component of Tris-glycine and Tris-glycine-SDS running buffers for polyacrylamide gel electrophoresis. Glycine is also a component of Towbin's transfer buffer for Western blots.


Inhibitory neurotransmitter in spinal cord, allosteric regulator of NMDA receptors.

Formula: NH₂CH₂COOH
Boiling Pt: >233 °C (dec.)
Melting Pt: 240 °C
Density: 1.16 g/mL
Storage Temperature: Ambient
MDL Number: MFCD00008131
CAS Number: 56-40-6
EINECS: 200-272-2
Merck Index: 13,04504
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d944007d30d5d4650062014a1b152bd9bfb66ea3&vid=093f4227cbdd1c4c6515ef9c4b85d93799877dcc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Identity Test Passes
Purity ≥99.5%
FTIR Conforms to Standard
Loss on Drying ≤0.2%
Residue on Ignition ≤.10%
Heavy Metals ≤20 ppm
Chloride ≤0.007%
Sulfate ≤0.0065%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR