Glycine ≥99.5%, white crystalline powder for electrophoresis
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Synonyms:
Aminoacetic acid, Glycocoll, H-Gly-OH
Glycine is an non-essential amino acid with no asymmetric carbon and major inhibitory neurotransmitter. It can fit into hydrophilic or hydrophobic environments, due to its minimal side chain of only one hydrogen atom. It contains heavy metals ≤ 20 ppm. It is commonly used as a component in Tris-glycine and Tris-glycine-SDS running buffers for polyacrylamide gel electrophoresis, a component of Towbin's transfer buffer for Western blots, a buffer substance in cryoenzymology, in osmotic pressure maintenance in isoelectric focusing of erythrocytes, salting-in effect in protein chemistry, and as a buffer component in the coupled phosphatase-kinase reaction for end labelling of restriction fragments.
Glycine is a component of Tris-glycine and Tris-glycine-SDS running buffers for polyacrylamide gel electrophoresis. Glycine is also a component of Towbin's transfer buffer for Western blots.
Inhibitory neurotransmitter in spinal cord, allosteric regulator of NMDA receptors.
- Electrophoresis Grade
- Presentation: White Crystalline Powder
- pKa at 25°C=2.35
- Soluble in water, slightly soluble in alcohol and ethanol(95%); almost insoluble in ether.
- Room Temperature
Glycine is an non-essential amino acid with no asymmetric carbon and major inhibitory neurotransmitter. It can fit into hydrophilic or hydrophobic environments, due to its minimal side chain of only one hydrogen atom. It contains heavy metals ≤ 20 ppm. It is commonly used as a component in Tris-glycine and Tris-glycine-SDS running buffers for polyacrylamide gel electrophoresis, a component of Towbin's transfer buffer for Western blots, a buffer substance in cryoenzymology, in osmotic pressure maintenance in isoelectric focusing of erythrocytes, salting-in effect in protein chemistry, and as a buffer component in the coupled phosphatase-kinase reaction for end labelling of restriction fragments.
Glycine is a component of Tris-glycine and Tris-glycine-SDS running buffers for polyacrylamide gel electrophoresis. Glycine is also a component of Towbin's transfer buffer for Western blots.
Inhibitory neurotransmitter in spinal cord, allosteric regulator of NMDA receptors.
Formula:
NH₂CH₂COOH Boiling Pt: >233 °C (dec.) Melting Pt: 240 °C Density: 1.16 g/mL Storage Temperature: Ambient |
MDL Number:
MFCD00008131 CAS Number: 56-40-6 EINECS: 200-272-2 Merck Index: 13,04504 |
|
Specification Test Results
Identity Test | Passes |
Purity | ≥99.5% |
FTIR | Conforms to Standard |
Loss on Drying | ≤0.2% |
Residue on Ignition | ≤.10% |
Heavy Metals | ≤20 ppm |
Chloride | ≤0.007% |
Sulfate | ≤0.0065% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...