3-[(3-Cholamidopropyl)dimethylammonio]-1-propane sulfate (CHAPS) ≥98%, white powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: 3-[(3-Cholamidopropyl)dimethylammonio]-1-propanesulfonate, 3-[(3-Cholamidopropyl)dimethylammonio]-1-propanesulphonate, CHAPS

04808196
IC808196EA 180.31 USD
IC808196
3-[(3-Cholamidopropyl)dimethylammonio]-1-propane sulfate (CHAPS) ≥98%, white powder
3-[(3-Cholamidopropyl)dimethylammonio]-1-propane sulphate (CHAPS)

CHAPS is useful for solubilizing membrane proteins. Frequently used prior to electrophoresis since it generally does not denature the proteins.


Soluble in 10% aqueous solution.(Lit.)


CHAPS is useful for solubilizing membrane proteins. Frequently used prior to electrophoresis since it generally does not denature the proteins.


Caution: Research Use Only (RUO).

Formula: C₃₂H₅₈N₂O₇S
MW: 614.89 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00081083
CAS Number: 75621-03-3

Order Now

Specification Test Results

Identity Test Passes
Purity ≥98%
FTIR Conforms to Standard
OD 400 (5% aq.) 0.03
OD 290 (5% aq.) 0.10

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR