Bromophenol blue, powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: Bromphenol Blue Sultone Form, 3,3',5,5'-Tetrabromophenolsulfonphthalein, BPB, 3,3',5,5'-Tetrabromophenolsulphonphthalein, Tetrabromophenol-sulphonephthalein, Tetrabromophenol-sulfonephthalein

04805732
IC805732EA 54.04 USD
IC805732
Bromophenol blue, powder
Bromophenol blue

Soluble in ethanol (0.1% w/v-clear yellow solution), methanol (40mg/4mL), EGME, water, benzene.


Visual Transition Interval: Yellow (pH 3.0) to Purple (pH 4.6)


Bromophenol blue migrates at approximately the same rate as 300-500bp DNA in agarose gel and at the buffer front in protein polyacrylamide gels.


Bromophenol blue is a tracking dye for alkaline and neutral buffer systems. It is used as a tracking dye in DNA, RNA (agarose) and protein (polyacrylamide) gel electrophoresis.


Visual Transition Interval: Yellow (pH 3.0) to Purple (pH 4.6)


Bromophenol blue migrates at approximately the same rate as 300-500bp DNA in agarose gel and at the buffer front in protein polyacrylamide gels.

Formula: C₁₉H₁₀Br₄O₅S
MW: 669.97 g/mol
Melting Pt: 273 °C (lit.)
MDL Number: MFCD00005875
CAS Number: 115-39-9
EINECS: 204-086-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR