Citric acid, anhydrous ≥99.5%, crystalline powder for electrophoresis

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: 2-Hydroxypropane-1,2,3-tricarboxylic acid, Hydroxytricarballylic acid

04800681
IC800681EA 142.35 USD
IC800681
Citric acid, anhydrous ≥99.5%, crystalline powder for electrophoresis
Citric acid

Soluble in water(50 g/mL).


Citric acid acts as an iron chelator. It can support cholesterol/sterol, ubiquinone and isoprenoid biosynthesis in cell culture applications. In animals, citric acid improves the utilization of nutritional calcium. It is a useful buffer component.


Citric acid acts as an iron chelator. It can support cholesterol/sterol, ubiquinone and isoprenoid biosynthesis in cell culture applications. In animals, citric acid improves the utilization of nutritional calcium. It is a useful buffer component.

Formula: HOC(COOH)(CH₂COOH)₂
MW: 192.13 g/mol
Boiling Pt: 310
MDL Number: MFCD00011669
CAS Number: 77-92-9
Merck Index: 13,02350
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d60f5a9b8e6c6dfb7eda04e1a1d5d6d971f92444&vid=caf4f9effc7fd22b942f7f92be770b06e40637a4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Synonyms β-Hydroxytricarballylic Acid; Citreoviridin
Form White Crystalline Powder
Storage Store at Room Temperature(15- 30 °C).
Heavy Metals 10 ppm
pH 1 - 3 (5% aq soln) (Lit.)
pka (1) 3.13, (2) 4.76, (3) 6.4
Residue on Ignition 0.05%
Water Content 0.5%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR