Plate Sealer, Acetate, Non-Sterile, MP Biomedicals
Supplier: MP Biomedicals
097640105
IC7640105CS 84.29 USD
IC7640105
Plate Sealer, Acetate, Non-Sterile, MP Biomedicals
Seals Microplate Seals
This microplate sealing tape is suitable for all microplates for both short- and long-term storage, providing a tight seal to minimize evaporation and condensation.
  • Easy application and removal for all microplates
  • Both short- and long-term storage
  • Suitable for use between -16 °C and 38 °C
  • Transparent, not pierceable, nonsterile
Used for sealing multi-well plates and tissue culture plates.