ImmunO™ Sheep Serum, MP Biomedicals
Supplier: MP Biomedicals
08642951
IC642951EA 146.83 USD
IC642951
ImmunO™ Sheep Serum, MP Biomedicals
Sera (Serum)
Sheep serum is useful for the propagation of long-term cultures of erythrocytic stages of the Theileria uilenbergi intracellular protozoan parasite.
  • Sterile filtered
Reconstitute with 50 mL of sterile distilled water.