ImmunO™ Rat Serum, MP Biomedicals
Supplier: MP Biomedicals
08642941
IC642941EA 140.28 USD
IC642941
ImmunO™ Rat Serum, MP Biomedicals
Sera (Serum)
Normal rat serum may be used as a blocking agent and controls in immunoassays
  • Sterile filtered
When reconstituted with 10 mL (sterile) distilled water or buffer, as appropriate for application, protein concentration will be at 5.5 - 7.0 gm/dL.
This rat serum is pooled from a normal donor population.