ImmunO™ Goat Serum, MP Biomedicals
Supplier: MP Biomedicals
08642921
IC642921EA 337.9 USD
IC642921
ImmunO™ Goat Serum, MP Biomedicals
Sera (Serum)
Goat serum can be used in immunocytochemistry and blocking.
  • Sterile filtered