Rabbit Complement Type (HLA-DR), MP Biomedicals
Supplier: MP Biomedicals
08642812
IC642812EA 231.02 USD
IC642812
Rabbit Complement Type (HLA-DR), MP Biomedicals
Sera (Serum)
The complement is produced from a large pool of New Zealand White rabbits, 8 - 12 weeks old
Reconstitute the lyophilized serum with an equal amount of sterile distilled water. After reconstitution store at 2 - 8 °C and use within eight hours. Discard any unused thawed complement after eight hours.
The animals are of United States origin. Cytotoxic titer is determined by NIH standards for microlymphocytotoxicity. Minimum release criteria is 80 - 100% cell death in 1:2 dilution.