Cell Biology Murashige and Skoog Modified Vitamin Solution (1000X), MP Biomedicals
Supplier: 0000012207
Contains the vitamins as described by Murashige and Skoog (1962) modified with additional Thiamine.
- Plant Media
- Typical Working Concentration: 1 mL/ L of Medium
- Soluble in water
Formula for solution (mg/L): Glycine (Free Base): 2000, myo-Inositol: 100,000, Nicotinic Acid (Free Acid): 500, PyridoxineHCI: 500, ThiamineHCI: 1000
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...