Cell Biology Murashige and Skoog Medium, without Surcrose, Agar, IAA and Kinetin, MP Biomedicals
Supplier: 0000012207
092610020 092610022 092610024
IC2610020CS 84.58 USD
IC2610020 IC2610022 IC2610024
Cell Biology Murashige and Skoog Medium, without Surcrose, Agar, IAA and Kinetin, MP Biomedicals
Media for Microbiology Dehydrated Media for Microbiology
Murashige and Skoog Medium contains Murashige and Skoog salts, as well as a mixture of vitamins
  • Plant Media
  • Effective in the culture of tissue derived from moncotyledons and dicotyledons
  • Batches are checked to ensure they conform to preset high standards
Formulation: Component: Amino Acids-Glycine: 2.00000 mg/L; Vitamins- myo-Inositol: 100.00000 mg/L; Niacin: 0.50000 mg/L; Pyridoxine HCl: 0.50000 mg/L; Thiamine HCl 0.10000 mg/L; Inorganic Salts-Ammonium Nitrate [NH4NO3]: 1650.00000 mg/L; Boric Acid [H3BO3]: 6.20000 mg/L; Calcium Chloride [CaCl2 2H2O] (anhyd): 332.0000 mg/L; Cobalt Chloride [CoCl2 6H2O] Hexahydrate: 0.02500 mg/L; Cupric Sulfate [CuSO4]: 0.01600 mg/L; EDTA Fe Na: 36.70000 mg/L; Magnesium Sulfate [MgSO4]: 180.70000 mg/L; Manganese Sulfate [MnSO4 H2O] Monohydrate: 16.90000 mg/L; Potassium Iodide [KI]: 0.83000 mg/L; Potassium Nitrate [KNO3]: 1900.00000 mg/L; Potassium Phosphate Monobasic [KH2PO4]: 170.00000 mg/L; Sodium Molybdate [Na2MoO4 2H2O] Dihydrate: 0.25000 mg/L; Zinc Sulfate [ZnSO4 7H2O] Heptahydrate: 8.60000 mg/L
This modification of the original formula does not contain agar, sucrose, IAA, and Kinetin.