Acid red 94, dark red powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

0219549105 0219549125
IC19549105EA 96.05 USD
IC19549105 IC19549125
Acid red 94, dark red powder
Rose Bengal

Rose Bengal is a fluorescent probe for determining NAD binding sites of alcohol dehydrogenase.


Used in Conn′s technique for bacteria in soil; as an alternative to phloxine B in Kreyberg′s stain for keratin and mucus.


Soluble in water at 1 mg/mL (pink to red solution).(Lit.)


Rose Bengal is a fluorescent probe for determining NAD binding sites of alcohol dehydrogenase.

Formula: C₂₀H₂Cl₄I₄O₅Na₂
Storage Temperature: Ambient
MDL Number: MFCD00151169
CAS Number: 632-69-9

Order Now

Specification Test Results

Identity Test Passes
UV/Vis: λmax (water) Store at Room Temperature (15 to 30 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR