Potassium dihydrogen phosphate, anhydrous ≥99.0%, crystalline powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

02195453.1 02195453.5 0219545301
IC195453.1EA 66.17 USD
IC195453.1 IC195453.5 IC19545301
Potassium dihydrogen phosphate, anhydrous ≥99.0%, crystalline powder
Potassium dihydrogen phosphate

Potassium phosphate is used as a component for a wide variety of media used in the culture of microorganisms, as a component in phosphate buffered saline (PBS). In addition to helping maintain pH, it supplies essential phosphate.


Soluble in water; insoluble to slightly soluble in ethanol; insoluble in alcohol.


Caution: Research Use Only (RUO).

Formula: KH₂PO₄
MDL Number: MFCD00011401
CAS Number: 7778-77-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fece579009266617bdb412ca5cac027e06c5836e&vid=fece579009266617bdb412ca5cac027e06c5836e&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=49fb72b1e887b0a1196b74f441a13bf74710f4e9&vid=49fb72b1e887b0a1196b74f441a13bf74710f4e9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Identity Test Passes
Assay ≥99.0%
pH (5% aq soln) 4.1 - 4.5


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR