Carbenicillin disodium salt, free flowing powder

Supplier: 0000012207
Danger

02195092.2 0219509201 0219509205
IC195092.2EA 74.86 USD
IC195092.2 IC19509201 IC19509205
Carbenicillin disodium salt, free flowing powder
Carbenicillin disodium salt

Semi-synthetic antibiotic related to penicillin. It is a benzyl penicillin derivative with substitution of a-carboxyl group n the benzyl side chain. The product interferes with final cell wall synthesis of susceptible bacteria. It is completely ionized in the blood. Carbencillin disodium is about 50% reversibly protein bound in plasma and its tissue distribution is similar to other penicillins.


  • Presentation: Free Flowing Powder
  • pKa: (in water) 3.3
  • Soluble in water (50 mg/ ml) and alcohol


Though Carbencillin disodium has substantial in vitro activity against a variety of both gram-positive and gram-negative microorganisms, the most important aspect of its profile is in its antipseudomonal and antiproteal effect.


Used for selection of ampr transformed cells and to study the role of penicillin-sensitive transpeptidases in cell wall biosynthesis.


Carboxypenicillin antibiotic that inhibits bacterial cell-wall synthesis (peptidoglycan cross-linking) by inactivating transpeptidases on the inner surface of the bacterial cell membrane. Analog to ampicillin. Antimicrobial spectrum: Gram-positive and Gram-negative bacteria, Pseudomonas.

Formula: C₁₇H₁₆N₂O₆S·2Na
Storage Temperature: Refrigerator
MDL Number: MFCD00077683
CAS Number: 4800-94-6

Order Now

Specification Test Results

Identity Test Passes
Potency 650 µg/mg


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR