Lipoprotein-Cholesterol concentrate, MP Biomedicals
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Lipoprotein-cholesterol is a mixture of lipoprotein cholesterol, phospholipids and fatty acids.
- Filtered through 0.2 μm membrane filter
- No preservatives
- Free of enzyme inhibitors
- From Bovine Serum
- Cholesterol concentration: 1000-2000 mg/dL
For use as a serum-free or serum-reduced media nutrient supplement, calibration of cholesterol values, and enzymatic or non-enzymatic cholesterol determinations.