Sodium hydrogen carbonate 99.7-100.3% (dried basis), white powder ACS

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: Sodium bicarbonate

02191435.5
IC191435.5EA 72.08 USD
IC191435.5
Sodium hydrogen carbonate 99.7-100.3% (dried basis), white powder ACS
Sodium hydrogen carbonate

Soluble in Water.


Chloride (Cl) 0.003%; Phosphate (PO4) 0.001%; Sulfur Compounds (as SO4) 0.003%; Ammonium (NH4) 5 ppm; Magnesium (Mg) 0.005%; Potassium (K) 0.005%; Calcium (Ca) 0.02%; Iron (Fe) 0.001%

Formula: NaHCO₃
MW: 84.01 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003528
CAS Number: 144-55-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f6b1b80d2afb3deab0c4ca836a921fcaad637890&vid=1163b37d9cf1c10fe0ed252e02a51fe12d6b50dd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Identity Test Passes
Assay 99.7 - 100.3% (dried basis)
Insoluble Matter ≤0.015%
Chloride (Cl) 0.003%
Phosphate (PO4) 0.001%
Sulphur Compounds (as SO4) 0.003%
Ammonium (NH4) 5 ppm
Magnesium (Mg) 0.005%
Potassium (K) 0.005%
Calcium (Ca) 0.02%
Heavy Metals (as Pb) ≤5 ppm
Iron (Fe) 0.001%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR