Rabbit Serum, MP Biomedicals
Supplier: MP Biomedicals
0219135780
IC19135720EA 51.46 USD
IC19135720 IC19135780
Rabbit Serum, MP Biomedicals
Sera (Serum)
Rabbit serum is a component of blood obtained from a rabbit that contains blood plasma without fibrinogen or other clotting factors
  • Total Protein= 4.0 - 7.0 gm/dL
  • pH: 6.5 - 8.0
  • Presentation: Frozen Brown Solution
In immunohistochemistry, antibodies are used to detect the presence and abundance of certain antigens in a given tissue sample. The use of a blocking serum such as rabbit serum is important because it prevents nonspecific binding and interaction between the antibodies and non-targeted antigens. Rabbit Serum is also a common component in media used to maintain cell culture.
Rabbit Serum Combine with BSA to use as a blocking serum in immunocytochemistry and immunohistochemistry
Total Protein= 4.0 - 7.0 gm/dL
Rabbit serum is a component of blood obtained from a rabbit that contains blood plasma without fibrinogen or other clotting factors.