Guanine, white powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: 2-Amino-1H-purin-6(7H)-one

0219069125
IC19069125EA 111.68 USD
IC19069125
Guanine, white powder
Guanine

Guanine is one of the four main nucleobases found in the nucleic acids DNA and RNA. Guanine is a purine derivative.


Guanine is used as an additive to various products (e.g., shampoos), where it provides a pearly iridescent effect. It is also used in metallic paints and simulated pearls and plastics. It provides shimmering luster to eye shadow and nail polish.


Soluble in 0.1 N HCl (50 mg/mL - clear, colorless solution), 1 M Sodium hydroxide (50 mg/mL - clear to slightly hazy light yellow solution). Freely soluble in ammonia water, aqueous KOH solutions, dilute acids; sparingly soluble in alcohol, ether. Almost insoluble in water.


Guanine is used as an additive to various products (e.g., shampoos), where it provides a pearly iridescent effect. It is also used in metallic paints and simulated pearls and plastics. It provides shimmering luster to eye shadow and nail polish.

Formula: C₅H₅N₅O
MW: 151.13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00071533
CAS Number: 73-40-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR