Sodium dodecyl sulfate (SDS) ≥95%
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Soluble in water (200 mg/mL - clear, faint yellow solution), and ethanol (0.1g/10 mL)
SDS is an detergent typically used to solubilize and denature proteins for electrophoresis. SDS has also been used in large-scale phenol extraction of RNA to promote the dissociation of protein from nucleic acids when extracting from biological material. SDS is used in Protein Electrophoresis & Blotting.
Formula:
CH₃(CH₂)₁₁OSO₃Na |
MDL Number:
MFCD00036175 CAS Number: 151-21-3 UN: 1325 ADR: 4.1,II |
|
Specification Test Results
Synonyms | Sodium Lauryl Sulfate; Sulfuric Acid, Monododecyl Ester, Sodium Salt; Dodecyl Sulfate, sodium salt; SLS; SDS; Neutrazyme; Lauryl Sulfate Sodium Salt; SDS Running Buffer |
Form | White Powder |
Storage | Store at Room Temperature (15-30 °C). |
Aggregation Number | 62 |
CMC | 8.27 mM |
pH | 6 - 7 (1% aq soln) |
Refractive Index | 1.461(Lit.) |
Typical Working Concentration |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...