Lidocaine, white powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Warning

0219011125 0219011183
IC19011125EA 72.08 USD
IC19011125 IC19011183
Lidocaine, white powder
Lidocaine

Na+ channel blocker; class IB antiarrhythmic that is rapidly absorbed after parenteral administration.


Lidocaine is used to treat allergies. It is generally used to differentiate and identify leucocytes, malaria parasites, and trypanosomas.


Soluble to 100 mM in ethanol.


Lidocaine is used to treat allergies. It is generally used to differentiate and identify leucocytes, malaria parasites, and trypanosomas.


Caution: Research Use Only (RUO).

Formula: C₁₄H₂₂N₂O
Melting Pt: 68,5 °C
Storage Temperature: Ambient
MDL Number: MFCD00026733
CAS Number: 137-58-6

Order Now

Specification Test Results

Identity Test Conforms to Standard


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR