Dextran sulfate sodium salt, M.W. ∼36000 - 50000, white powder, Colitis Grade

Supplier: 0000012207

0216011080 0216011001 0216011010 0216011050 0216011090 0216011025 0216011091
IC16011080EA 2506.99 USD
IC16011080 IC16011001 IC16011010 IC16011050 IC16011090 76202-276 76203-054
Dextran sulfate sodium salt, M.W. ∼36000 - 50000, white powder, Colitis Grade
Dextran sulphate sodium salt

Dextran sulfate is a polyanionic derivative of dextran produced by esterification of Dextran with chlorosulphonic acid. Dextran sulfate sodium (DSS) is the sodium salt which is a white to off-white powder freely soluble in water and salt solutions to form a stable, clear solution. The sulfur content is approximately 17% which corresponds to an average of 1.9 sulfate groups per glucosyl residue of the dextran molecule.


Dextran Sulfate Sodium (DSS) which has proven to have versatile value in Biomedical/clinical research. DSS can be used to study Colitis (Inflammatory Bowel disease) as well as an antiviral as agent.


Inflammatory Bowel Disease (IBD) is characterized by chronic and relapsing inflammation of the gastrointestinal tract which is associated with increased risk of developing colitis-associated cancer. Several animal models have been used to study colitis and one such model involves the oral administration of dextran sodium sulfate (DSS) in the drinking water of mice leading to chronic colitis. This DSS induced colitis model is spontaneous and is used to assess the therapeutic potential of treatments for IBD.

Formula: (C₆H₇Na₃O₁₄S₃)n
Storage Temperature: Ambient
MDL Number: MFCD00081551
CAS Number: 9011-18-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=70dadbbe11c99e497a247142bfc87b7d1b5703d3&vid=70dadbbe11c99e497a247142bfc87b7d1b5703d3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Identity Test Passes
Moisture 10%
Residue on Ignition 50%
Sulfur ~18 - 20%
Glucose 35 - 50%
pH (1% aq soln) 5 - 7.5
Specific Rotation +75° to +105°
Free Sulfate ≤0.2%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR