Daidzein ≥98%, powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: Daidzeol, 7-Hydroxy-3-(4-hydroxyphenyl)-4-benzopyrone, 7-Hydroxy-3-(4-hydroxyphenyl)chromen-4-one, 4',7-Dihydroxyisoflavone, 7-Hydroxy-3-(4-hydroxyphenyl)-4H-chromen-4-one

0215881280
IC15881280EA 512.01 USD
IC15881280
Daidzein ≥98%, powder
Daidzein

Daidzein is a negative control for MP Genistein (tyrosine kinase inhibitor, Cat. No. 152355).


Daidzein is a phytoestrogen that is suggested to play a role in preventing hormone-induced cancers. Arrests cell cycle at G1 in Swiss 3T3 cells. Soy isoflavone daidzein protects against oxidative damage in liver cells induced by 7,12 - dimethylbenz[a]anthracene (DMBA). Catalase and superoxide dismutase activity, down-regulated by DMBA, was restored by daidzein.


Soluble in DMSO.


Daidzein is a negative control for MP Genistein (tyrosine kinase inhibitor, Cat. No. 152355).

Formula: C₁₅H₁₀O₄
MW: 254.24 g/mol
Boiling Pt: 513
MDL Number: MFCD00016954
CAS Number: 486-66-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR