Fluorescent brightener 28, powder microbiology stain
Supplier: 0000012207
Fluorescent brightener 28 is used for the staining of fungi and as a viability stain.
- Colourless organic compound that fluoresces (clear bluish) under ultraviolet radiation
- Working Concentration 0.05–0.20%
- Related structurally to Fluorescent Brightener 9 (Calcofluor White MR)
- Originally used in plant tissue studies
Industrial uses as a fluorescent brightening agent for cellulose and polyamide fabrics, paper, in detergents and soaps.
This product is related structurally to Fluorescent Brightner 9 (Calcofluor White MR), which was used by Jefferies and Belcher and by Maeda and Ischida in plant tissue studies.
Fluorescent Brightener 28 is used for the staining of fungi and as a viability stain. Industrial uses include use as a fluorescent brightening agent for cellulose and polyamide fabrics, paper and in detergents and soaps.
Caution: In case of contact with eyes, rinse immediately with plenty of water and seek medical advice.
Formula:
C₄₀H₄₄N₁₂O₁₀S₂ |
MDL Number:
MFCD00308955 CAS Number: 4404-43-7 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...