Iron(II) sulfate heptahydrate ≥99%, bluish green granular powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Warning

Synonyms: Iron vitriol, Ferrous sulphate heptahydrate, Ferrous sulfate heptahydrate

0215804883
IC15804883EA 81.4 USD
IC15804883
Iron(II) sulfate heptahydrate ≥99%, bluish green granular powder
Iron (II) sulphate heptahydrate

Ferrous sulphate heptahydrate used in the manufacturing of iron compounds, in electroplating baths, aluminium etching, process engraving and lithography, and in redox polymerisation. It is used as a reducing agent in chemical processes, leather dyes and writing inks.


Also, it is a component of weed killers, wood preservatives and water treatment processes. It is used in prevention of chlorosis in plants and therapeutically as hematinic. In veterinary medicine, it is used as an astringent to combat iron deficiency.


Soluble in water (100 mg/mL); practically insoluble in alcohol.


Ferrous sulfate heptahydrate used in the manufacturing of iron compounds, in electroplating baths, aluminium etching, process engraving and lithography, and in redox polymerisation. It is used as a reducing agent in chemical processes, leather dyes and writing inks. Also, it is a component of weed killers, wood preservatives and water treatment processes. It is used in prevention of chlorosis in plants and therapeutically as hematinic. In veterinary medicine, it is used as an astringent to combat iron deficiency.

Formula: FeSO₄·7H₂O
MW: 278.02 g/mol
MDL Number: MFCD00149719
CAS Number: 7782-63-0

Order Now

Specification Test Results

Identity Test Passes
Purity ≥99%
FTIR Conforms to Reference

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR