Iron(III) sulfate hydrate, pale gray powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Warning

0215804283 0215804290 0215804291
IC15804283EA 81.86 USD
IC15804283 IC15804290 IC15804291
Iron(III) sulfate hydrate, pale gray powder
Iron (III) sulphate hydrate

Ferric Sulfate is used in dyeing as a mordant, and as a coagulant for industrial wastes. It is also used in pigments, and in pickling baths for aluminum and steel. Medically it is used as an astringent and styptic. It also acts as an important oxidative agent.


Soluble in water; sparingly soluble in alcohol; practically insoluble in acetone, ethyl acetate.


Ferric Sulfate is used in dyeing as a mordant, and as a coagulant for industrial wastes. It is also used in pigments, and in pickling baths for aluminum and steel. Medically it is used as an astringent and styptic. It also acts as an important oxidative agent.

Formula: Fe₂(SO₄)₃·xH₂O
Melting Pt: 1539 °C (lit.)
Storage Temperature: Ambient
MDL Number: MFCD00149714
CAS Number: 15244-10-7

Order Now

Specification Test Results

Identity Test Passes
Moisture Record Result


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR