Ethoxyquin ≥90%, viscous liquid

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: 6-Ethoxy-1,2-dihydro-2,2,4-trimethylquinoline, 6-Ethoxy-2,2,4-trimethyl-1,2-dihydroquinoline, 1,2-Dihydro-6-ethoxy-2,2,4-trimethylquinoline, Ethoxyquin

0215796380
IC15796380EA 129.49 USD
IC15796380
Ethoxyquin ≥90%, viscous liquid
Ethoxyquin

Ethoxyquin is a quinoline-based antioxidant.


Ethoxyquin is an antioxidant which inhibits chemically induced carcinogenesis, where it exerts its chemopreventive action by inducing glutathione synthetase and the phase II drug metabolizing enzyme, glutathione S-transferase (GST) and an antidegradation agent for rubber.


Ethoxyquin is an antioxidant which inhibits chemically induced carcinogenesis, where it exerts its chemopreventive action by inducing glutathione synthetase and the phase II drug metabolizing enzyme, glutathione S-transferase (GST) and an antidegradation agent for rubber.

Formula: C₁₄H₁₉NO
MDL Number: MFCD00023883
CAS Number: 91-53-2

Order Now

Specification Test Results

Identity Test Passes
Purity ≥90%
FTIR Conforms to Standard

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR