Methyl ethyl ketone, liquid

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: EK 500 Additive, Butan-2-one, Ethyl methyl ketone, 2-Butanone, Butanone, MEK

0215554891
IC15554891EA 106.26 USD
IC15554891
Methyl ethyl ketone, liquid
Methyl ethyl ketone

Soluble in alcohol, ether and most fixed oils. Soluble in water.

Formula: C₂H₅COCH₃
MW: 72.11 g/mol
MDL Number: MFCD00011648
CAS Number: 78-93-3
UN: 1193
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8b6656888f6a9a4c0c5ed3053bb2a3dcd8b3af66&vid=17cb98abf677b90f8bac752664d5c0cfd6765591&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR