Methyl cellulose, white powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

0215549283 0215549291
IC15549283EA 199.37 USD
IC15549283 IC15549291
Methyl cellulose, white powder
Methyl cellulose

Methyl cellulose is a chemical compound derived from cellulose. It is a hydrophilic white powder in pure form and dissolves in cold (but not in hot) water, forming a clear viscous solution or gel. It is used as a thickener and emulsifier in various food and cosmetic products. It is not digestible, not toxic, and not an allergen.


Methyl cellulose is used as a substitute for water-soluble gums,to render paper greaseproof in adhesives,as thickening agent in cosmetics,as protective colloid in emulsions,as binder and stabilizer in foods,as bulk producer in the formulation of dietetic foods,Pharmaceutic aid (suspending agent),Laxative.


Soluble in cold water. Insoluble in hot water.


Prepared from wood pulp by treatment with alkali and methylation of the alkali cellulose with methyl chloride.


Methoxy Groups-27.5 - 31.5%


Methyl cellulose is a chemical compound derived from cellulose. It is a hydrophilic white powder in pure form and dissolves in cold (but not in hot) water, forming a clear viscous solution or gel. It is used as a thickener and emulsifier in various food and cosmetic products. It is not digestible, not toxic, and not an allergen.

Storage Temperature: Ambient
MDL Number: MFCD00081763
CAS Number: 9004-67-5

Order Now

Specification Test Results

Identity Test Passes
Viscosity (2% aqueous solution at 25 °C) ~15 centipoises
Loss on Drying ≤5.0%
Methoxy groups 27.5 - 31.5%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR