2-Bromethylamine hydrobromide ≥98.0%, white crystalline powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Warning

Synonyms: 2-Bromoethylammonium bromide, 2-Bromoethylamine hydrobromide

0215487525
IC15487525EA 45.21 USD
IC15487525
2-Bromethylamine hydrobromide ≥98.0%, white crystalline powder
2-Bromethylamine hydrobromide

2-Bromoethylamine is used to construct C2-symmetric imidazolidinylidene ligands with a dioxolane backbone.


2-Bromoethylamine is used to construct C2-symmetric imidazolidinylidene ligands with a dioxolane backbone.


Caution: Research Use Only (RUO).

Formula: BrCH₂CH₂NH₃Br
MW: 204.89 g/mol
Melting Pt: 342…345° F (lit.)
Storage Temperature: Ambient
MDL Number: MFCD00012886
CAS Number: 2576-47-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=92954711ef75996bd496abcb054ff7876386defe&vid=92954711ef75996bd496abcb054ff7876386defe&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR