5-Aminosalicylic acid ≥99%, powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: Mesalamine, Mesalazine, 5-Amino-2-hydroxybenzoic acid, m-Aminosalicylic acid

0215477025 0215477080
IC15477025EA 341.63 USD
IC15477025 IC15477080
5-Aminosalicylic acid ≥99%, powder
5-Aminosalicylic acid

5-Aminosalicylic acid is a peroxidase substrate suitable for use in ELISA procedures. This substrate produces a soluble end product that is brown in color and can be read spectrophotometrically at 450 nm. The reaction may be stopped with 3 N NaOH and read at 550 nm .


Solubility: 200 mg plus 4 ml of 1 M HCl. Slightly soluble in cold water and alcohol. More soluble in hot water.


5-Aminosalicylic acid is a peroxidase substrate suitable for use in ELISA procedures. This substrate produces a soluble end product that is brown in color and can be read spectrophotometrically at 450 nm. The reaction may be stopped with 3 N NaOH and read at 550 nm .

Formula: C₇H₇NO₃
MW: 153.14 g/mol
Boiling Pt: 347
Melting Pt: 280
MDL Number: MFCD00007877
CAS Number: 89-57-6
Merck Index: 12,05964

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR