Calcium chloride, anhydrous ≥95%, white powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Warning

Synonyms: Chloro calcium, Calcium dichloride, CaCl2

0215350280 0215350290
IC15350280EA 57.44 USD
IC15350280 IC15350290
Calcium chloride, anhydrous ≥95%, white powder
Calcium chloride

Anhydrous Calcium Chloride is highly hygroscopic and is often used as a desiccant. Calcium plays important roles in many biological processes, including signal transduction, muscle contraction, maintenance of cell membranes and cell wall stability.


Soluble in water with heat liberation, ethanol, acetic acid, acetone.

Formula: CaCl₂
Storage Temperature: Ambient
MDL Number: MFCD00010903
CAS Number: 10043-52-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e966cfdab71ec84617dd9126afc84e397bdacf4c&vid=e966cfdab71ec84617dd9126afc84e397bdacf4c&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR