Sodium hydroxide ≥97%, pellets

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: Caustic soda, Caustic soda, Sodium hydrate, Lye, Soda lye

02153495.5 0215349501
IC153495.5EA 95.93 USD
IC153495.5 IC15349501
Sodium hydroxide ≥97%, pellets
Sodium hydroxide

1g dissolves in 0.9 mL water; 0.3 mL boiling water, 7.2 mL absolute alcohol, 4.2 mL methanol; insoluble in glycerol. The dissolution of sodium hydroxide in water is a highly exothermic reaction. Sodium hydroxide stock solutions should be stored in plastic containers. Glass containers should be avoided in the preparation and storage of sodium hydroxide solutions because it will etch the glass (typically anything over 2 N will etch glass). Keep solutions protected from air as much as possible since contact with air will form sodium carbonate.


Poison/Deleterious Substance Code: 2-54

Formula: NaOH
MDL Number: MFCD00003548
CAS Number: 1310-73-2
UN: 1823
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6a6113fc30d522dfb01b99ca89f67657aac15cbc&vid=6a6113fc30d522dfb01b99ca89f67657aac15cbc&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR