Strontium chloride hexahydrate 99.0-103.0%, crystalline powder ACS

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

0215258380
IC15258380EA 76.38 USD
IC15258380
Strontium chloride hexahydrate 99.0-103.0%, crystalline powder ACS
Strontium chloride hexahydrate

Sulfate NMT 0.001% ;Barium NMT 0.05% ; Calcium NMT 0.05% ; Magnesium NMT0.0002%

Formula: SrCl₂·6H₂O
MDL Number: MFCD00149865
CAS Number: 10025-70-4

Order Now

Specification Test Results

Synonyms Strontium Dichloride Hexahydrate
Form White, Hard Packed, Crystalline Powder
Storage Store at Room Temperature (15-30 °C).
Assay 99.0-103.0%
Heavy Metals NMT 0.0005% ; Iron NMT 0.0005%
Insoluble Matter NMT 0.005%
pH 5.0-7.0 (5% sol)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR