Salicylic acid ≥99.0%, white powder ACS

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: 2-Hydroxybenzoic acid, o-Hydroxybenzoic acid

0215257080 0215257090
IC15257080EA 88.71 USD
IC15257080 IC15257090
Salicylic acid ≥99.0%, white powder ACS
Salicylic acid

Salicylic acid is a natural product extract from Willow bark, well known as an antiinflammatory and antinociceptive agent and a close structural relative to acetylsalicylic acid (aspirin).


Salicylic acid is a ubiquitous plant hormone with many regulatory functions involved in local disease resistance mechanisms and systemic acquired resistance. It is also of interest as a starting material for the organic synthesis of more elaborate COX suppressors and other chemical structures.


The anti-inflammatory and antinociceptive effects produced by salicylic acid and its derivatives in animals are due to inhibition of COX-1 and COX-2 (cyclooxygenase) enzyme activity and suppression of prostaglandin biosynthesis.


Chloride (CI): 0.001% ; Sulfate (SO4): ≤0.003%


Salicylic acid is a ubiquitous plant hormone with many regulatory functions involved in local disease resistance mechanisms and systemic acquired resistance. It is also of interest as a starting material for the organic synthesis of more elaborate COX suppressors and other chemical structures.

Formula: (HO)C₆H₄CO₂H
Storage Temperature: Ambient
MDL Number: MFCD00002439
CAS Number: 69-72-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=af1f695fa92656a135b5e1742f1cca28f4d255fb&vid=8f3aff6fb9a70e5dd194b8bc46019af74e48e90a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99,0%
Heavy Metals as Pb ≤5 ppm;
Melting Point 158.0 - 161.0°C
Residue on Ignition ≤0,01%
Chloride 0,001%
Sulfate (as SO₄) ≤0,003%
Iron (Fe) ≤2 ppm
Substances Darkened by Sulfuric Acid Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR