Potassium thiocyanate ≥98.6%, powder ACS
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
0215256680
0215256690
IC15256680EA
107.75
USD
IC15256680
IC15256690
Potassium thiocyanate ≥98.6%, powder ACS
Potassium thiocyanate
Specification Test Results
Assay | 98.64% |
pH (5% soln) | 6.84 |
Insoluble in Water | 0.00% |
Chloride (Cl) |
|
Sulfate (SO4) |
|
Ammonium (NH4) |
|
Heavy Metals (as Pb) |
|
Iron (Fe) |
|
Sodium (Na) | 0.00% |
Iodine Consuming Substances | Passes |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...