Molybdenum(VI) oxide, powder ACS
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
|
Warning
|
Molybdenum Trioxide is used as a precursor to LAMOX fast ion conductors and superconductors. Also Used in the solid state synthesis of a remarkable ternary, reduced molybdenum oxide, Pr4Mo9O18, whose structure contains previously unknown Mo7, Mo13 and Mo19 clusters. The new cluster product is a small band gap semiconductor.
Solubility: Less than 1 mg/mL H₂O at 75 °F (NTP, 1992)
Caution: Research Use Only (RUO).
Formula:
MoO₃ |
MDL Number:
MFCD00003469 CAS Number: 1313-27-5 UN: 3288 ADR: 6.1,III |
Specification Test Results
Assay | ≥99,5% |
Insoluble in Dilute Ammonium Hydroxide | ≤0,01% |
Chloride (Cl) | ≤0,002% |
Nitrate | Passes |
Arsenate, Phosphate, and Silicate | ≤0,001% |
Phosphate | ≤5 ppm |
Sulfate | ≤0,02% |
Ammonium | ≤0,002% |
Heavy Metals (Pb) | ≤0,005% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...