Molybdenum(VI) oxide, powder ACS

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Warning

0215254880 0215254890
IC15254880EA 112.97 USD
IC15254880 IC15254890
Molybdenum(VI) oxide, powder ACS
Molybdenum (VI) oxide

Molybdenum Trioxide is used as a precursor to LAMOX fast ion conductors and superconductors. Also Used in the solid state synthesis of a remarkable ternary, reduced molybdenum oxide, Pr4Mo9O18, whose structure contains previously unknown Mo7, Mo13 and Mo19 clusters. The new cluster product is a small band gap semiconductor.


Solubility: Less than 1 mg/mL H₂O at 75 °F (NTP, 1992)


Caution: Research Use Only (RUO).

Formula: MoO₃
MDL Number: MFCD00003469
CAS Number: 1313-27-5
UN: 3288
ADR: 6.1,III

Order Now

Specification Test Results

Assay ≥99,5%
Insoluble in Dilute Ammonium Hydroxide ≤0,01%
Chloride (Cl) ≤0,002%
Nitrate Passes
Arsenate, Phosphate, and Silicate ≤0,001%
Phosphate ≤5 ppm
Sulfate ≤0,02%
Ammonium ≤0,002%
Heavy Metals (Pb) ≤0,005%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR