D-(-)-Mannitol ≥99.0%, white powder ACS

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: Mannitol, (2R,3R,4R,5R)-Hexane-1,2,3,4,5,6-hexaol

0215254083 0215254090 0215254091
IC15254083EA 93.34 USD
IC15254083 IC15254090 IC15254091
D-(-)-Mannitol ≥99.0%, white powder ACS
D(-)-Mannitol

A carbohydrate found in plants and plant exudates.


Used with boric acid in the manufacturing of dry electrolytic condensers for radio applications.


Soluble in water (1 g/5.5 mL), alcohol (1 g/83 mL), glycerol (1 g/18 mL); more soluble in hot water; soluble in pyridine, aniline, and aqueous solutions of alkalies; insoluble in ether.


Caution: Research Use Only (RUO).

Formula: HOH₂C(CH(OH))₄CH₂OH
MW: 182.17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00064287
CAS Number: 69-65-8
Merck Index: 13,05769
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=163f556dca45086e05ff490ddc8eeb65d04aea05&vid=408e3f5566e309064ce68988cdd160305c5b8c0a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity ≥99.0%
Optical Rotation +23.3° to +24.3°
Reducing Sugars Passes
Insoluble Matter ≤0.01%
Loss on Drying ≤0.05%
Residue After Ignition ≤0.01%
Titratable Acid 0.0008 meq/g
Heavy Metals (as Pb) ≤5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR