Copper(I) chloride ≥90.0%, powder ACS

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: Cuprous chloride, Copper chloride

0215251580
IC15251580EA 162.91 USD
IC15251580
Copper(I) chloride ≥90.0%, powder ACS
Copper (I) chloride

Insoluble in Acid: 0.02%, Sulfate: 0.1%, Sodium: 0.05%, Potassium: 0.02%, Calcium: 0.01%

Formula: ClCu
MW: 99 g/mol
Boiling Pt: 1367
Melting Pt: 430
MDL Number: MFCD00010971
CAS Number: 7758-89-6
UN: 2802
ADR: 8 III
Merck Index: 13,02690

Order Now

Specification Test Results

Synonyms Copper(I) chloride; Copper monochloride; Cuprous chloride
Form Lime Green Powder
Storage Store at Room Temperature (15-30 °C). Store Desiccated. Protect from light.
Assay ≥90.0%
Heavy Metals Iron: ≤0.005%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR