Hexachloroplatinic(IV) acid hexahydrate (37,50% Pt), powder ACS

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Danger

Synonyms: Hydrogen hexachloroplatinate (IV) hexahydrate, Chloroplatinic acid hexahydrate, Dihydrogen hexachloroplatinate (IV) hexahydrate

0215250901 0215250905
IC15250901EA 207.64 USD
IC15250901 IC15250905
Hexachloroplatinic(IV) acid hexahydrate (37,50% Pt), powder ACS
Hexachloroplatinic (IV) acid hexahydrate

Chloroplatinic acid is used in potassium determination, purification of platinum and in catalysis.


Soluble in alcohol; very soluble in water.


Alkali & Other Salts (as Sulfates) ≤0.05%


Chloroplatinic acid is used in potassium determination, purification of platinum and in catalysis.

Formula: H₂PtCl₆·6H₂O
Melting Pt: 60 °C (lit.)
MDL Number: MFCD00149910
CAS Number: 18497-13-7
UN: 2507
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9e367658e5dd86f9c4a8b84cf3fa0199847c0881&vid=9e367658e5dd86f9c4a8b84cf3fa0199847c0881&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR