Ammonium heptamolybdate tetrahydrate, white crystalline powder ACS

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Warning

0215249780 0215249790 0215249791
IC15249780EA 111.96 USD
IC15249780 IC15249790 IC15249791
Ammonium heptamolybdate tetrahydrate, white crystalline powder ACS
Ammonium heptamolybdate tetrahydrate

Ammonium Molybdate Tetrahydrate is an analgesic.


Ammonium Molybdate Tetrahydrate is an analgesic.


Caution: Research Use Only (RUO).

Formula: (NH₄)₆Mo₇O₂₄·4H₂O
Storage Temperature: Ambient
MDL Number: MFCD00167059
CAS Number: 12054-85-2

Order Now

Specification Test Results

Assay (as MoO3) 81,0 - 83,0%
Heavy Metals (as Pb) ≤0,001%
Insoluble Matter ≤0,005%
Chloride (Cl) ≤0,002%
Arsenate, Phosphate and Silicate (as SiO2) ≤0,001%
Phosphate (PO4) ≤5 ppm
Sulphate (SO4) ≤0,02%
Magnesium ≤,005%
Potassium ≤0,002%
Sodium ≤0,01%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR