Ammonium heptamolybdate tetrahydrate, white crystalline powder ACS
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
|
Warning
|
0215249780
0215249790
0215249791
IC15249780EA
111.96
USD
IC15249780
IC15249790
IC15249791
Ammonium heptamolybdate tetrahydrate, white crystalline powder ACS
Ammonium heptamolybdate tetrahydrate
Ammonium Molybdate Tetrahydrate is an analgesic.
Ammonium Molybdate Tetrahydrate is an analgesic.
Caution: Research Use Only (RUO).
Formula:
(NH₄)₆Mo₇O₂₄·4H₂O Storage Temperature: Ambient |
MDL Number:
MFCD00167059 CAS Number: 12054-85-2 |
Specification Test Results
Assay | (as MoO3) 81,0 - 83,0% |
Heavy Metals | (as Pb) ≤0,001% |
Insoluble Matter | ≤0,005% |
Chloride (Cl) | ≤0,002% |
Arsenate, Phosphate and Silicate (as SiO2) | ≤0,001% |
Phosphate (PO4) | ≤5 ppm |
Sulphate (SO4) | ≤0,02% |
Magnesium | ≤,005% |
Potassium | ≤0,002% |
Sodium | ≤0,01% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...