Genistein, free flowing white powder
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
|
Warning
|
Synonyms:
5,7-Dihydroxy-3-(4-hydroxyphenyl)-4-benzopyrone, 5,7-Dihydroxy-3-(4-hydroxyphenyl)-4H-1-benzopyran-4-one, 5,7-Dihydroxy-3-(4-hydroxyphenyl)-4H-chromen-4-one, 4',6'-Dihydroxy-3-(4-hydroxyphenyl)benzo(e)oxacyclohex-2-en-4-on, 5,7-Dihydroxy-3-(4-hydroxyphenyl)chromen-4-one, 4',5,7-Trihydroxyisoflavone
Genistein inhibits EGF-stimulated phosphorylation in cultured cells. Also used to explore signal transduction pathways.
- Inhibitor of tyrosine protein kinase
- Competitive inhibitor of ATP in other protein kinase reactions Antiangiogenic agent, down-regulates the transcription of genes involved in controlling angiogenesis
Soluble in water above pH 8 (at 40 micrograms/ml), DMSO at 50 mg/ml, and in DMF. Practically insoluble in cold water; slightly soluble in hot water, hot ethanol, and hot methanol and soluble in hot 80% ethanol, hot 80% methanol, hot acetone, and pyridine. Warning: alkaline hydrolysis yields phloroglucinal and p-hydroxyphenyl-acetic acid.
ID50 for EGF receptor kinase = 0.7 ug/mL.
Caution: Research Use Only (RUO).
Formula:
C₁₅H₁₀O₅ Melting Pt: 301,5 °C (dec.) |
MDL Number:
MFCD00016952 CAS Number: 446-72-0 |
Specification Test Results
Identity Test | Passes |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...