Genistein, free flowing white powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Warning

0215235510 0215235550
IC15235510EA 81.92 USD
IC15235510 IC15235550
Genistein, free flowing white powder
Genistein

Genistein inhibits EGF-stimulated phosphorylation in cultured cells. Also used to explore signal transduction pathways.


  • Inhibitor of tyrosine protein kinase
  • Competitive inhibitor of ATP in other protein kinase reactions Antiangiogenic agent, down-regulates the transcription of genes involved in controlling angiogenesis


Soluble in water above pH 8 (at 40 micrograms/ml), DMSO at 50 mg/ml, and in DMF. Practically insoluble in cold water; slightly soluble in hot water, hot ethanol, and hot methanol and soluble in hot 80% ethanol, hot 80% methanol, hot acetone, and pyridine. Warning: alkaline hydrolysis yields phloroglucinal and p-hydroxyphenyl-acetic acid.


ID50 for EGF receptor kinase = 0.7 ug/mL.


Caution: Research Use Only (RUO).

Formula: C₁₅H₁₀O₅
Melting Pt: 301,5 °C (dec.)
MDL Number: MFCD00016952
CAS Number: 446-72-0

Order Now

Specification Test Results

Identity Test Passes


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR