Tris(hydroxymethyl)aminomethane (TRIS, Trometamol) ≥99.0%, Reagent Grade

Supplier: 0000012207

02152176.1 02152176.5 0215217601 0215217605 0215217610
IC152176.1EA 44.7 USD
IC152176.1 IC152176.5 IC15217601 IC15217605 IC15217610
Tris(hydroxymethyl)aminomethane (TRIS, Trometamol) ≥99.0%, Reagent Grade
Tris(hydroxymethyl)aminomethane (TRIS, Trometamol)

40% in water.

Formula: NH₂C(CH₂OH)₃
MDL Number: MFCD00004679
CAS Number: 77-86-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6afa5ccd09fa1d4cc16871d24bacf8028a3ec1f1&vid=6afa5ccd09fa1d4cc16871d24bacf8028a3ec1f1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Synonyms Trometamol; Tromethamine; Tris-[hydroxymethyl]aminomethane; 2-Amino-2-(hydroxymethyl)-1,3-propanediol
Form White Crystalline Powder
Storage Store at Room Temperature (15 - 30 °C), desiccate.
pH 10 - 11.5 (5% aq soln)
pka 8.1 (25 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR