Safranine O, dark green powder C.I. 50240
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
|
Warning
|
Safranine O is a redox and adsorption indicator.
Safranine O is a biological stain used in histology, cytology, bacteriology.
Soluble in ethanol 50% (Clear, orange-red solution with fluorescence).
Formula:
C₂₀H₁₉ClN₄ Storage Temperature: Ambient |
MDL Number:
MFCD00011759 CAS Number: 477-73-6 |
Specification Test Results
Identity Test | Passes |
UV/Vis: l max (50% ethanol) | 530 ± 5 nm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...