Propionic acid ≥98.5%, liquid

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: Propanoic acid

0215195680
IC15195680EA 46.56 USD
IC15195680
Propionic acid ≥98.5%, liquid
Propionic acid
Formula: C₃H₆O₂
MW: 74.08 g/mol
MDL Number: MFCD00002756
CAS Number: 79-09-4
UN: 3463
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR