1-Hexanesulfonic acid sodium salt, white flakes

Supplier: MP BIOMEDICALS (FKA ICN BIOMED

Synonyms: Sodium hexane-1-sulfonate, Sodium hexane-1-sulphonate, Sodium hexanesulfonate, Sodium hexanesulphonate, Sodium 1-hexanesulfonate, Sodium 1-hexanesulphonate

0215125210
IC15125210EA 87.59 USD
IC15125210 76102-716
1-Hexanesulfonic acid sodium salt, white flakes
1-Hexanesulphonic acid sodium salt

Sodium hexanesulfonate is a low molecular weight alkylsulfonate that is used as an ion pairing reagent for HPLC and as an anionic surfactant. The anionic sulfonate counterion permits the separation and resolution of positively charged analytes. Because of its low molecular weight, it will not form micelles in solution and therefore has no CMC value.
For ion-pair chromatography. Sodium hexanesulfonate is utilized in the analysis of small organic compounds, and pharmaceutical products and metabolites by such techniques as HPLC.


  • Presentation: White Flakes
  • Soluble in water (100 mg/ml-clear, colorless), ethanol or methanol
  • Storage temperature: Room Temperature

Formula: C₆H₁₃O₃SNa
MW: 188.22 g/mol
Melting Pt: 300
Storage Temperature: Ambient
MDL Number: MFCD00007542
CAS Number: 2832-45-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=aebc3cb7a7cefac9ad5a7678d82a90d244b508f9&vid=d608216fe1e698d6b206b03588e6d6fe5ee661e1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Identity Test Passes
FTIR Conforms to Reference
Moisture ≤10%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR